N-Succinimidyl Ferrocenecarboxylate ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: Ferrocenecarboxylic Acid N-Succinimidyl Ester

S0820-1G S0820-200MG
TCIAS0820-1GEA 3075 NOK
TCIAS0820-1G TCIAS0820-200MG
N-Succinimidyl Ferrocenecarboxylate ≥98.0% (by HPLC)
N-Succinimidyl Ferrocenecarboxylate
Formel: C₁₅H₁₃FeNO₄
MW: 327.12 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD03427488
CAS-nummer: 115223-09-1

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Chelometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR