SIA (N-Succinimidyl iodoacetate) ≥95.0% (by total nitrogen basis)

Leverandør: TCI
Warning

Synonymer: SIA

S0844-100MG
TCIAS0844-100MGEA 1800 NOK
TCIAS0844-100MG
SIA (N-Succinimidyl iodoacetate) ≥95.0% (by total nitrogen basis)
SIA (N-Succinimidyl iodoacetate)
Formel: C₆H₆INO₄
MW: 283.02 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00058451
CAS-nummer: 39028-27-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (with Total Nitrogen) min. 95.0 %
Melting point 146.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR