Sodium stearoyllactate (so called)
Leverandør: TCI
Synonymer: Stearoyllactic Acid Sodium Salt
S0846-25G
TCIAS0846-25GEA 220 NOK
TCIAS0846-25G
Sodium stearoyllactate (so called)
Sodium stearoyllactate
Bestill nå
Spesifikasjon Testresultater
Appearance White to Orange to Green powder to crystal
Ester Value 150 to 190
Acid Value 60 to 80
Heavy metals (as Pb) max. 10 ppm
Arsenic (as As2O3) max. 1.0 ppm