N-Succinimidyl Bromoacetate ≥98.0% (by GC)

Leverandør: TCI
Warning

S0852-100MG
TCIAS0852-100MGEA 650 NOK
TCIAS0852-100MG
N-Succinimidyl Bromoacetate ≥98.0% (by GC)
N-Succinimidyl Bromoacetate
Formel: C₆H₆BrNO₄
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00058571
CAS-nummer: 42014-51-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 110.0 to 118.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR