SATP (N-Succinimidyl 3-(Acetylthio)propionate) ≥95.0% (by GC)

Leverandør: TCI

Synonymer: SATP

S0859-100MG
TCIAS0859-100MGEA 1450 NOK
TCIAS0859-100MG
SATP (N-Succinimidyl 3-(Acetylthio)propionate) ≥95.0% (by GC)
SATP (N-Succinimidyl 3-(Acetylthio)propionate)
Formel: C₉H₁₁NO₅S
MW: 245.25 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00133023
CAS-nummer: 84271-78-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc548afe5b7dc044905c4b56be4c235490201df&vid=1bc548afe5b7dc044905c4b56be4c235490201df&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 64.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR