N-Succinimidyl 5-Azido-2-nitrobenzoate ≥97.0% (by HPLC)

Leverandør: TCI

Synonymer: 5-Azido-2-nitrobenzoic Acid N-Succinimidyl Ester, ANB-NOS, N-(5-Azido-2-nitrobenzoyloxy)succinimide

S0860-10MG
TCIAS0860-10MGEA 635 NOK
TCIAS0860-10MG
N-Succinimidyl 5-Azido-2-nitrobenzoate ≥97.0% (by HPLC)
N-Succinimidyl 5-Azido-2-nitrobenzoate
Formel: C₁₁H₇N₅O₆
MW: 305.21 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00054962
CAS-nummer: 60117-35-3

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Nitrogen) 22.20 to 23.20 %
Melting point 133.0 to 137.0 deg-C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR