SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate) ≥97.0% (by HPLC)

Leverandør: TCI
Warning

S0861-10MG
TCIAS0861-10MGEA 515 NOK
TCIAS0861-10MG
SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate) ≥97.0% (by HPLC)
SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate)
Formel: C₁₈H₁₆N₂O₆
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00054961
CAS-nummer: 79886-55-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 129.0 to 133.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR