D-erythro-Sphingosine ≥96.0% (by GC)

Leverandør: TCI

Synonymer: D-Sphingosine, trans-D-erythro-2-Amino-4-octadecene-1,3-diol, (2S,3R,E)-2-Amino-4-octadecene-1,3-diol, 4-Sphingenine, (2S,3R,E)-2-Aminooctadec-4-ene-1,3-diol

S0874-25MG
TCIAS0874-25MGEA 1450 NOK
TCIAS0874-25MG
D-erythro-Sphingosine ≥96.0% (by GC)
D-erythro-Sphingosine
Formel: C₁₈H₃₇NO₂
MW: 299.50 g/mol
Smeltepunkt: 74°C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00036751
CAS-nummer: 123-78-4
EINECS: 204-651-3
Merck Indeks: 14,08747
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3c98c16d0a8931c50af353b61a78e3e824724fc&vid=b3c98c16d0a8931c50af353b61a78e3e824724fc&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Specific rotation [a]20/D -0.9 to -3.0 deg (C=1, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR