N-Succinimidyl-11-maleimidoundecanoate ≥98.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: N-(11-Maleimidoundecanoyloxy)succinimide, KMUS, 11-Maleimidoundecanoic Acid N-Succinimidyl Ester

S0882-20MG
TCIAS0882-20MGEA 1375 NOK
TCIAS0882-20MG
N-Succinimidyl-11-maleimidoundecanoate ≥98.0% (by HPLC)
N-Succinimidyl-11-maleimidoundecanoate
Formel: C₁₉H₂₆N₂O₆
MW: 378.43 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00467873
CAS-nummer: 87981-04-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42acdcb0fccfd5c08972c3302c39741536dba677&vid=42acdcb0fccfd5c08972c3302c39741536dba677&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 7.1 to 7.5 %
Melting point 90.0 to 94.0 deg-C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR