Sodium-5-(2-aminoethylamino)-1-naphthalenesulphonate hydrate ≥98.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: Sodium N-(2-Aminoethyl)-5-naphthylamine-1-sulphonate, 5-(2-Aminoethylamino)-1-naphthalenesulphonic acid Sodium Salt, N-(2-Aminoethyl)-5-naphthylamine-1-sulphonic acid Sodium Salt, 1,5-EDANS sodium salt, EDANS, sodium salt

S0891-100MG
TCIAS0891-100MGEA 445 NOK
TCIAS0891-100MG
Sodium-5-(2-aminoethylamino)-1-naphthalenesulphonate hydrate ≥98.0% (by HPLC)
Sodium-5-(2-aminoethylamino)-1-naphthalenesulphonate hydrate
Formel: C₁₂H₁₄N₂O₃S
MW: 288.30 g/mol
MDL nummer: MFCD00051474
CAS-nummer: 100900-07-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76842593788a72f113732719ef64f54a17a07ad5&vid=1eb716453a30baa2d7f9ec8a0000bca5f348ceb1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 % (calcd.on anh.substance)
Water max. 14.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR