N-Succinimidyl 4-Formylbenzoate ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: Ald-Ph-NHS ester, 2,5-Dioxopyrrolidin-1-yl 4-formylbenzoate, 4-Formylbenzoic Acid N-Succinimidyl Ester

S0893-100MG
TCIAS0893-100MGEA 2925 NOK
TCIAS0893-100MG
N-Succinimidyl 4-Formylbenzoate ≥98.0% (by HPLC)
N-Succinimidyl 4-Formylbenzoate
Formel: C₁₂H₉NO₅
MW: 247.21 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00133733
CAS-nummer: 60444-78-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85558f4f9da41d0e6b104099069a5d0c0ae44ca2&vid=85558f4f9da41d0e6b104099069a5d0c0ae44ca2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 168.0 to 175.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR