N-Succinimidyl trifluoroacetate ≥95.0% (by GC)

Leverandør: TCI
Warning

Synonymer: TFA-NHS, N-(Trifluoroacetoxy)succinimide, Trifluoroacetic acid N-Succinimidyl ester, 2,5-Dioxopyrrolidin-1-yl 2,2,2-trifluoroacetate

S0915-1G
TCIAS0915-1GEA 875 NOK
TCIAS0915-1G
N-Succinimidyl trifluoroacetate ≥95.0% (by GC)
N-Succinimidyl trifluoroacetate
Formel: C₆H₄F₃NO₄
MW: 211.10 g/mol
Kokepunkt: 100°C
MDL nummer: MFCD00595252
CAS-nummer: 5672-89-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54bbd510ddeb4d2e64b24e06db983494da9fc857&vid=54bbd510ddeb4d2e64b24e06db983494da9fc857&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR