(+)-Dimethyl-L-tartrate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: Dimethyl (2R,3R)-2,3-dihydroxysuccinate, (R,R)-(+)-Tartaric acid dimethyl ester, (R,R)-(+)-Dimethyl tartrate, L(+)-Tartaric acid dimethyl ester, (+)-Dimethyl L-tartrate

T0006-25G
TCIAT0006-25GEA 300 NOK
TCIAT0006-25G
(+)-Dimethyl-L-tartrate ≥98.0% (by GC)
(+)-Dimethyl-L-tartrate
Formel: C₆H₁₀O₆
Kokepunkt: 118°C
Smeltepunkt: 58…61 °C
Tetthet: 1,3403 g/cm³ (15 °C)
MDL nummer: MFCD00064437
CAS-nummer: 608-68-4
EINECS: 210-166-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C
Specific rotation [a]20/D +20.0 to +22.0 deg (C=1, H2O)
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR