Erythrosin B ≥95.0% (by HPLC)

Leverandør: TCI

Synonymer: C.I. 45430:2

T0124-25G
TCIAT0124-25GEA 1350 NOK
TCIAT0124-25G
Erythrosin B ≥95.0% (by HPLC)
Erythrosin B
Formel: C₂₀H₈I₄O₅
MW: 835.89 g/mol
MDL nummer: MFCD00005044
CAS-nummer: 15905-32-5
EINECS: 240-046-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af30453a3162a340c12c7482c7eb2519f63df355&vid=af30453a3162a340c12c7482c7eb2519f63df355&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 95.0 area%
Solubility in EtOH (95vol%) almost transparency
Sensitiveness to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR