1,3-Diphenyl-2-thiourea ≥98.0% (by total nitrogen basis)

Leverandør: TCI
Danger

T0197-25G T0197-500G
TCIAT0197-25GEA 220 NOK
TCIAT0197-25G TCIAT0197-500G
1,3-Diphenyl-2-thiourea ≥98.0% (by total nitrogen basis)
1,3-Diphenyl-2-thiourea
Formel: C₁₃H₁₂N₂S
Kokepunkt: 365 °C (1013 hPa)
Smeltepunkt: 150…153 °C
Tetthet: 1,3205 g/cm³ (20 °C)
MDL nummer: MFCD00004921
CAS-nummer: 102-08-9
EINECS: 203-004-2
FN: 2811
ADR: 6.1,II
Merck Indeks: 14,03333
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72105fe3d59860807259d820a2af527e117688c4&vid=72105fe3d59860807259d820a2af527e117688c4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 150.0 to 153.0 deg-C
Solubility in hot Methanol almost transparency

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR