m-Toluidin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

T0298-25ML T0298-500ML
TCIAT0298-25MLEA 245 NOK
TCIAT0298-25ML TCIAT0298-500ML
m-Toluidin ≥98.0% (by GC, titration analysis)
m-Toluidin
Formel: C₇H₉N
Kokepunkt: 203°C
Smeltepunkt: –30 °C
Tetthet: 0,98444 g/cm³ (20 °C)
Flash punkt: 86°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00007808
CAS-nummer: 108-44-1
EINECS: 203-583-1
FN: 1708
ADR: 6.1,II
Merck Indeks: 14,09536
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6e753bf6f27b453867b1daa903484c4e47b96f8&vid=c6e753bf6f27b453867b1daa903484c4e47b96f8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR