m-Toluidine hydrochloride 98,0 GC, N

Leverandør: TCI
Danger

Synonymer: M-toluidine hydrochloride

T0301-25G
TCIAT0301-25GEA 1925 NOK
TCIAT0301-25G
m-Toluidine hydrochloride 98,0 GC, N
m-Toluidine hydrochloride
Formel: C₇H₉N
MW: 143.61 g/mol
MDL nummer: MFCD00070607
CAS-nummer: 638-03-9
EINECS: 211-314-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR