p-Tolyltetrazolium red ≥95.0% (by HPLC)

Leverandør: TCI

Synonymer: 2,5-Diphenyl-3-(p-tolyl)tetrazolium Chloride

T0326-100MG
TCIAT0326-100MGEA 835 NOK
TCIAT0326-100MG
p-Tolyltetrazolium red ≥95.0% (by HPLC)
p-Tolyltetrazolium red
Formel: C₂₀H₁₇ClN₄
MW: 348.83 g/mol
MDL nummer: MFCD00060004
CAS-nummer: 71658-33-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR