1,3,3-Trimethylindolino-6'-bromobenzopyrylospiran ≥98.0% (by HPLC, titration analysis) photochromic compound

Leverandør: TCI

Synonymer: 6-Bromo-1',3',3'-trimethylspiro[1(2H)-benzopyran-2,2'-indoline], 6-Bromo-1',3',3'-trimethylspiro[chromene-2,2'-indoline]

T0370-1G
TCIAT0370-1GEA 1650 NOK
TCIAT0370-1G
1,3,3-Trimethylindolino-6'-bromobenzopyrylospiran ≥98.0% (by HPLC, titration analysis) photochromic compound
1,3,3-Trimethylindolino-6'-bromobenzopyrylospiran
Formel: C₁₉H₁₈BrNO
MW: 356.26 g/mol
MDL nummer: MFCD00059896
CAS-nummer: 16650-14-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR