2-Tridekanon ≥95.0% (by GC)

Leverandør: TCI
Warning

T0415-25G
TCIAT0415-25GEA 580 NOK
TCIAT0415-25G
2-Tridekanon ≥95.0% (by GC)
2-Tridekanon
Formel: C₁₃H₂₆O
MW: 198,35 g/mol
Kokepunkt: 263°C
Smeltepunkt: 25…27 °C
Tetthet: 0,82 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008968
CAS-nummer: 593-08-8
EINECS: 209-784-0
FN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c22406aeed9cce74725f8e50c745a2da10d915e&vid=0c22406aeed9cce74725f8e50c745a2da10d915e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR