Trypan blue

Leverandør: TCI
Warning

Synonymer: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)

T0556-25G
TCIAT0556-25GEA 380 NOK
TCIAT0556-25G
Trypan blue
Trypan blue
Formel: C₃₄H₂₄N₆Na₄O₁₄S₄
MDL nummer: MFCD00003969
CAS-nummer: 72-57-1
EINECS: 200-786-7
Merck Indeks: 14,09792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f756e316d391053c75aa4592bce9ae1fa724569f&vid=f756e316d391053c75aa4592bce9ae1fa724569f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Dark green to Dark blue to Black powder to crystal
Lambda max. 594.0 to 602.0 nm (H2O)
Absorbance (E1%1cm) min. 600 (H2O, 594.0 to 602.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR