cis-3-Hexen-1-yltiglate ≥97.0% (by GC)

Leverandør: TCI
Warning

T0934-25ML
TCIAT0934-25MLEA 905 NOK
TCIAT0934-25ML
cis-3-Hexen-1-yltiglate ≥97.0% (by GC)
cis-3-Hexen-1-yltiglate
Formel: C₁₁H₁₈O₂
Flash punkt: 96°C
MDL nummer: MFCD00036528
CAS-nummer: 67883-79-8
EINECS: 267-554-5

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR