Tris(4-nitrophenyl)phosphate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

T1002-25G
TCIAT1002-25GEA 1325 NOK
TCIAT1002-25G
Tris(4-nitrophenyl)phosphate ≥98.0% (by HPLC, titration analysis)
Tris(4-nitrophenyl)phosphate
Formel: C₁₈H₁₂N₃O₁₀P
MDL nummer: MFCD00024649
CAS-nummer: 3871-20-3
EINECS: 223-389-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 154.0 to 159.0 deg-C
Solubility in hot Chloroform almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR