1,2,4-Trimetoksybensen ≥97.0% (by GC)

Leverandør: TCI

T1130-25G
TCIAT1130-25GEA 745 NOK
TCIAT1130-25G
1,2,4-Trimetoksybensen ≥97.0% (by GC)
1,2,4-Trimetoksybensen
Formel: C₉H₁₂O₃
MW: 168,19 g/mol
Kokepunkt: 247°C
Smeltepunkt: 19…21 °C
Tetthet: 1,126 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008360
CAS-nummer: 135-77-3
EINECS: 205-219-7
Merck Indeks: 13,01073
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97927e4404e548e7e6c9f19de7b2ee892b8a01b8&vid=97927e4404e548e7e6c9f19de7b2ee892b8a01b8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR