2,2':5',2''-Terthiophene ≥98.0% (by GC), purified by sublimation

Leverandør: TCI

T1196-1G T1196-5G
TCIAT1196-1GEA 245 NOK
TCIAT1196-1G TCIAT1196-5G
2,2':5',2''-Terthiophene ≥98.0% (by GC), purified by sublimation
2,2':5',2''-Terthiophene
Formel: C₁₂H₈S₃
MDL nummer: MFCD00012167
CAS-nummer: 1081-34-1
Merck Indeks: 14,09174

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Green powder to crystaline
Purity (GC) min. 98.0 %
Melting point 93.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR