1,3,3-Trimethylindolinonaphthospirooxazine ≥98.0% (by HPLC, titration analysis) photochromic compound

Leverandør: TCI
Warning

Synonymer: 1,3,3-Trimethylspiro[indoline-2,3'-[3H]naphth[2,1-b][1,4]oxazine], 1,3,3-Trimethylspiro[indoline-2,3'-naphtho[2,1-b][1,4]oxazine]

T1259-1G
TCIAT1259-1GEA 1450 NOK
TCIAT1259-1G
1,3,3-Trimethylindolinonaphthospirooxazine ≥98.0% (by HPLC, titration analysis) photochromic compound
1,3,3-Trimethylindolinonaphthospirooxazine
Formel: C₂₂H₂₀N₂O
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00013318
CAS-nummer: 27333-47-7
EINECS: 404-480-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR