Tetrabutylammonium bis(1,3-dithiole-2-thione-4,5-dithiolato)nickel(III)complex ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

T1272-100MG
TCIAT1272-100MGEA 540 NOK
TCIAT1272-100MG
Tetrabutylammonium bis(1,3-dithiole-2-thione-4,5-dithiolato)nickel(III)complex ≥98.0% (by titrimetric analysis)
Tetrabutylammonium bis(1,3-dithiole-2-thione-4,5-dithiolato)nickel(III)complex
Formel: C₂₂H₃₆NNiS₁₀
MW: 693,83 g/mol
MDL nummer: MFCD00059124
CAS-nummer: 68401-88-7

Bestill nå

Spesifikasjon Testresultater

Purity (HPLC) min. 97.0 area%
Purity (Chelometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR