Triethylamine phosphate 96,0 Titrimetric

Leverandør: TCI
Warning

Synonymer: Triethylammonium Phosphate

T1300-25G
TCIAT1300-25GEA 1575 NOK
TCIAT1300-25G
Triethylamine phosphate 96,0 Titrimetric
Triethylamine phosphate
Formel: C₆H₁₅N
MW: 199.19 g/mol
MDL nummer: MFCD00067475
CAS-nummer: 35365-94-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR