Trichloromethiazide ≥99.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: 6-Chloro-3-(dichloromethyl)-3,4-dihydro-2H-benzo[e][1,2,4]thiadiazine-7-sulphonamide 1,1-dioxide, 6-Chloro-3-dichloromethyl-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulphonamide 1,1-Dioxide

T1318-10G
TCIAT1318-10GEA 1600 NOK
TCIAT1318-10G
Trichloromethiazide ≥99.0% (by titrimetric analysis)
Trichloromethiazide
Formel: C₈H₈Cl₃N₃O₄S₂
MW: 380,64 g/mol
MDL nummer: MFCD00057315
CAS-nummer: 133-67-5
EINECS: 205-118-8
Merck Indeks: 14,09624

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 99.0 %
Solubility in Acetone almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR