1,6,20,25-Tetraaza[6.1.6.1]paracyclophane ≥98.0% (by HPLC)

Leverandør: TCI

T1378-100MG
TCIAT1378-100MGEA 1650 NOK
TCIAT1378-100MG
1,6,20,25-Tetraaza[6.1.6.1]paracyclophane ≥98.0% (by HPLC)
1,6,20,25-Tetraaza[6.1.6.1]paracyclophane
Formel: C₃₄H₄₀N₄
MW: 504.72 g/mol
MDL nummer: MFCD00142567
CAS-nummer: 74043-83-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR