Tetrabutylammonium bis(maleonitriledithiolato)nickel(III) complex ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Danger

T1415-1G
TCIAT1415-1GEA 2775 NOK
TCIAT1415-1G
Tetrabutylammonium bis(maleonitriledithiolato)nickel(III) complex ≥97.0% (by titrimetric analysis)
Tetrabutylammonium bis(maleonitriledithiolato)nickel(III) complex
Formel: C₂₄H₃₆N₅NiS₄
MDL nummer: MFCD00191492
CAS-nummer: 55401-12-2

Bestill nå

Spesifikasjon Testresultater

Appearance Black powder to crystal
Purity (Chelometric Titration) min. 97.0 %
Lambda max. 869.0 to 874.0 nm (CHCl3)
Molar extinction coefficient min. 900 (CHCl3, 869.0 to 874.0nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR