Thioindigo

Leverandør: TCI

Synonymer: Fluorescent Red Dye, Vat Red 41, 3H,3'H-[2,2'-Bibenzo[b]thiophenylidene]-3,3'-dione

T1489-25G
TCIAT1489-25GEA 715 NOK
TCIAT1489-25G
Thioindigo
Thioindigo
Formel: C₁₆H₈O₂S₂
MW: 296.36 g/mol
Smeltepunkt: 280°C
MDL nummer: MFCD00191692
CAS-nummer: 522-75-8
EINECS: 208-336-1

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown to Dark purple powder to crystal
Lambda max. 541.0 to 545.0 nm (Toluene)
Absorbance (E1%1cm) min. 450 (Toluene, 541.0 to 545.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR