L-(-)-Tryptophanol ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: L-Tryptophanol, (S)-(-)-2-Amino-3-(3-indolyl)propanol, (S)-2-Amino-3-(1H-indol-3-yl)propan-1-ol

T1537-1G
TCIAT1537-1GEA 1275 NOK
TCIAT1537-1G
L-(-)-Tryptophanol ≥97.0% (by titrimetric analysis)
L-(-)-Tryptophanol
Formel: C₁₁H₁₄N₂O
MW: 190,25 g/mol
MDL nummer: MFCD00037970
CAS-nummer: 2899-29-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D -19.0 to -22.0 deg (C=1, Methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR