1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-ɑ-D-galactopyranose

Leverandør: TCI

T1731-100MG
TCIAT1731-100MGEA 2375 NOK
TCIAT1731-100MG
1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-ɑ-D-galactopyranose
1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-alpha-D-galactopyranose
Formel: C₁₄H₁₉N₃O₉
MW: 373,32 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD01076182
CAS-nummer: 67817-30-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Melting point 120.0 to 127.0 deg-C
Specific rotation [a]20/D +98.0 to +102.0 deg (C=1, CHCl3)
Elemental analysis (Nitrogen) 11.00 to 11.40 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR