2,2':5',2''-Terthiophene-5-carboxaldehyde ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: 5-Formyl-2,2':5',2''-terthiophene, [2,2':5',2''-Terthiophene]-5-carbaldehyde

T1805-1G
TCIAT1805-1GEA 2175 NOK
TCIAT1805-1G
2,2':5',2''-Terthiophene-5-carboxaldehyde ≥98.0% (by HPLC)
2,2':5',2''-Terthiophene-5-carboxaldehyde
Formel: C₁₃H₈OS₃
MDL nummer: MFCD00115181
CAS-nummer: 7342-41-8

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 137.0 to 141.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR