Triphenylbismuth diacetate ≥98.0% (by titrimetric analysis)

Leverandør: TCI

T1836-5G
TCIAT1836-5GEA 2700 NOK
TCIAT1836-5G
Triphenylbismuth diacetate ≥98.0% (by titrimetric analysis)
Triphenylbismuth diacetate
Formel: C₂₂H₂₁BiO₄
MDL nummer: MFCD00092697
CAS-nummer: 7239-60-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c851ef0fd8058164d890954e5df1405b7bfd8a3c&vid=c851ef0fd8058164d890954e5df1405b7bfd8a3c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Solubility in hot Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR