2,6,6-Trimethyl-2,4-cycloheptadien-1-one ≥96.0% (by GC)

Leverandør: TCI

Synonymer: 2,6,6-Trimethylcyclohepta-2,4-dienone

T1846-5G
TCIAT1846-5GEA 3850 NOK
TCIAT1846-5G
2,6,6-Trimethyl-2,4-cycloheptadien-1-one ≥96.0% (by GC)
2,6,6-Trimethyl-2,4-cycloheptadien-1-one
Formel: C₁₀H₁₄O
MW: 150,22 g/mol
Kokepunkt: 82°C
MDL nummer: MFCD00182537
CAS-nummer: 503-93-5

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR