Triphenylantimony diacetate ≥98.0% (by HPLC)

Leverandør: TCI
Danger

Synonymer: Bis(acetato-O)triphenylantimony

T1848-1G
TCIAT1848-1GEA 690 NOK
TCIAT1848-1G
Triphenylantimony diacetate ≥98.0% (by HPLC)
Triphenylantimony diacetate
Formel: C₂₂H₂₁O₄Sb
MDL nummer: MFCD00127961
CAS-nummer: 1538-62-1
EINECS: 216-262-6
FN: 3467
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR