4-((Trifluoromethyl)thio)benzoic acid ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Warning

T2390-25G T2390-5G
TCIAT2390-25GEA 2550 NOK
TCIAT2390-25G TCIAT2390-5G
4-((Trifluoromethyl)thio)benzoic acid ≥97.0% (by GC, titration analysis)
4-((Trifluoromethyl)thio)benzoic acid
Formel: C₈H₅F₃O₂S
MDL nummer: MFCD00040906
CAS-nummer: 330-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1af8e1bef438d80aa8eb3ad982ed285e3bd464a4&vid=1af8e1bef438d80aa8eb3ad982ed285e3bd464a4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 159.0 to 163.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR