Tetrahydrofuran, vannfri ≥99.5% (by GC) stabilised

Leverandør: TCI
Danger


Stabilisering: stabilisert med BHT/Ionol

T2394-500ML
TCIAT2394-500MLEA 325 NOK
TCIAT2394-500ML
Tetrahydrofuran, vannfri ≥99.5% (by GC) stabilised
Tetrahydrofuran
Formel: C₄H₈O
Kokepunkt: 65°C
Smeltepunkt: –108°C
Tetthet: 0,888 g/cm³ (20 °C)
Flash punkt: –15°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005356
CAS-nummer: 109-99-9
EINECS: 203-726-8
FN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Indeks: 14,09211
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5878b108d8580b5945e2bc4a80d0feb033ea9423&vid=6aa88254eae0694713270f2c1f873b0d07a716ec&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 99.5 %
Water max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR