4-(Trifluoromethylthio)benzaldehyde ≥90.0% (by GC)

Leverandør: TCI
Warning

T2405-25G T2405-5G
TCIAT2405-25GEA 5900 NOK
TCIAT2405-25G TCIAT2405-5G
4-(Trifluoromethylthio)benzaldehyde ≥90.0% (by GC)
4-(Trifluoromethylthio)benzaldehyde
Formel: C₈H₅F₃OS
Kokepunkt: 85°C
MDL nummer: MFCD00129191
CAS-nummer: 4021-50-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57197bfaa8db4a3d1b1faa0984fbfce50bfc30fb&vid=57197bfaa8db4a3d1b1faa0984fbfce50bfc30fb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to lump
Purity (GC) min. 90.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR