2,2':5',2''-Terthiophene-5,5''-dicarboxaldehyde ≥98.0% (by GC)

Leverandør: TCI

T2639-1G
TCIAT2639-1GEA 3550 NOK
TCIAT2639-1G
2,2':5',2''-Terthiophene-5,5''-dicarboxaldehyde ≥98.0% (by GC)
2,2':5',2''-Terthiophene-5,5''-dicarboxaldehyde
Formel: C₁₄H₈O₂S₃
MDL nummer: MFCD01863637
CAS-nummer: 13130-50-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 220.0 to 224.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR