Tetradecanal ≥96.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Tetradecanaldehyde, Tetradecanal, Myristaldehyde, Tetradecyl aldehyde, Myristyl aldehyde

T2696-1G
TCIAT2696-1GEA 2650 NOK
TCIAT2696-1G
Tetradecanal ≥96.0% (by GC)
Tetradecanal
Formel: C₁₄H₂₈O
MW: 212,38 g/mol
Kokepunkt: 166°C
Smeltepunkt: 23°C
Tetthet: 0,83 g/cm³ (20 °C)
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00007019
CAS-nummer: 124-25-4
EINECS: 204-692-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea970b8a8fc662629e1fe65267238a9ff46d0499&vid=ea970b8a8fc662629e1fe65267238a9ff46d0499&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR