Tyrphostin AG 1478 ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: N-(3-Chlorophenyl)-6,7-dimethoxy-4-quinazolinamine, N-(3-Chlorophenyl)-6,7-dimethoxyquinazolin-4-amine

T2944-25MG
TCIAT2944-25MGEA 2400 NOK
TCIAT2944-25MG
Tyrphostin AG 1478 ≥98.0% (by HPLC)
Tyrphostin AG 1478
Formel: C₁₆H₁₄ClN₃O₂
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00270914
CAS-nummer: 153436-53-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bc1a8eca26f7d4d436276fdc6df995073c4d3d5&vid=7bc1a8eca26f7d4d436276fdc6df995073c4d3d5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR