Thio-NAD ≥90.0% (by HPLC) for biochemical research

Leverandør: TCI

Synonymer: (2R,3R,4S,5R)-5-[[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxymethyl]-2-(3-carbamothioylpyridin-1-ium-1-yl)-4-hydroxyoxolan-3-olate, Thionicotinamide-DPN, Thionicotinamide Adenine dinucleotide oxidized form

T2979-100MG
TCIAT2979-100MGEA 3150 NOK
TCIAT2979-100MG
Thio-NAD ≥90.0% (by HPLC) for biochemical research
Thio-NAD
Formel: C₂₁H₂₇N₇O₁₃P₂S
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00079609
CAS-nummer: 4090-29-3
EINECS: 223-828-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e5166a99b9eef6766d4c90f25da6819a6bab8c5&vid=6e5166a99b9eef6766d4c90f25da6819a6bab8c5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 90.0 area%
Functionality test to pass test

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR