1,3,3-Trimethyl-3,4-dihydroisoquinoline ≥98.0% (by GC, titration analysis)

Leverandør: TCI

Synonymer: 3,4-Dihydro-1,3,3-trimethylisoquinoline

T3060-1G
TCIAT3060-1GEA 1725 NOK
TCIAT3060-1G
1,3,3-Trimethyl-3,4-dihydroisoquinoline ≥98.0% (by GC, titration analysis)
1,3,3-Trimethyl-3,4-dihydroisoquinoline
Formel: C₁₂H₁₅N
Kokepunkt: 125°C
MDL nummer: MFCD00608546
CAS-nummer: 79023-51-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=957836ce779bbab50f2ebd1c3ad2d3a2d750bb16&vid=957836ce779bbab50f2ebd1c3ad2d3a2d750bb16&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light red clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR