Tetrabutylammonium bis(3,6-dichloro-1,2-benzenedithiolato)nickelate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: Tetrabutylammonium bis(3,6-dichloro-1,2-benzenedithiolato)nickel(III) Complex

T3204-200MG
TCIAT3204-200MGEA 1050 NOK
TCIAT3204-200MG
Tetrabutylammonium bis(3,6-dichloro-1,2-benzenedithiolato)nickelate ≥98.0% (by titrimetric analysis)
Tetrabutylammonium bis(3,6-dichloro-1,2-benzenedithiolato)nickelate
Formel: C₂₈H₄₀Cl₄NNiS₄
MW: 719,37 g/mol
MDL nummer: MFCD00274632
CAS-nummer: 87314-14-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9fc3d0d3b0539771c3fc338b0415863310fed8a&vid=a9fc3d0d3b0539771c3fc338b0415863310fed8a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Green to Dark green powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Melting point 203.0 to 207.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR