Tetrabutylammonium bis(3,4,6-trichloro-1,2-benzenedithiolato)nickelate ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: Tetrabutylammonium bis(3,4,6-trichloro-1,2-benzenedithiolato)nickel(III) Complex

T3205-200MG
TCIAT3205-200MGEA 970 NOK
TCIAT3205-200MG
Tetrabutylammonium bis(3,4,6-trichloro-1,2-benzenedithiolato)nickelate ≥95.0% (by titrimetric analysis)
Tetrabutylammonium bis(3,4,6-trichloro-1,2-benzenedithiolato)nickelate
Formel: C₂₈H₃₈Cl₆NNiS₄
MW: 788,25 g/mol
CAS-nummer: 87314-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acaae6480f1ed8735f90b697491cc0d39f31d6b0&vid=acaae6480f1ed8735f90b697491cc0d39f31d6b0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Green to Dark green powder to crystal
Purity (Chelometric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR