Tetrandine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: (4aS,16aS)-3,4,4a,5,16a,17,18,19-octahydro-12,21,22,26-tetramethoxy-4,17-dimethyl-16H-1,24:6,9-dietheno-11,15-metheno-2H-pyrido[2',3':17,18][1,11]dioxacycloeicosino[2,3,4-ij]isoquinoline, (1beta)-6,6',7,12-Tetramethoxy-2,2'-dimethylberbaman, (11S,31S)-16,36,37,54-Tetramethoxy-12,32-dimethyl-11,12,13,14,31,32,33,34-octahydro-2,6-dioxa-1(7,1),3(8,1)-diisoquinolina-5(1,3),7(1,4)-dibenzenacyclooctaphane, Tetrandrine, NSC 77037, d-Tetrandine

T3321-200MG T3321-1G
TCIAT3321-200MGEA 770 NOK
TCIAT3321-200MG TCIAT3321-1G
Tetrandine ≥98.0% (by HPLC, titration analysis)
Tetrandine
Formel: C₃₈H₄₂N₂O₆
MDL nummer: MFCD08689909
CAS-nummer: 518-34-3
Merck Indeks: 14,09231
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1a4b8d79b3688288e12d69d2d4988d0a59e73fc&vid=c1a4b8d79b3688288e12d69d2d4988d0a59e73fc&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 221.0 to 225.0 deg-C
Specific rotation [a]20/D +283 to +287 deg (C=1,CHCl3)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR