Trityl chloride resin 2,0-2,5 mmol/g, cross-linked with 1% DVB (200-400 mesh)
Leverandør: TCI
Warning
Synonymer: Trityl chloride, polymer-supported, Chlorodiphenylmethyl Polystyrene resin
T3432-1G
TCIAT3432-1GEA 905 NOK
TCIAT3432-1G
Trityl chloride resin 2,0-2,5 mmol/g, cross-linked with 1% DVB (200-400 mesh)
Trityl chloride resin
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5d6e42d4fdee14fa900516a1f128e5b26bd1c67&vid=ba74ec37b33b5452ec984965acb407d2e8e33a01&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Amber powder to crystal
Loading 2.0 to 2.5 mmol/g